• USA Home
  • Z282626 - Advanced Asymmetric Synthesis
EMAIL THIS PAGE TO A FRIEND
Z282626 Aldrich
Advanced Asymmetric Synthesis
  •  ISBN-10 0751400491
  •  ISBN-13 978-075140049-6
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   G.R. Stephenson, ed., Kluwer Academic Publishers, 1996, 442 pp., hard cover
mfr. no.   Kluwer Academic Publishers
Description
General description
Asymmetric synthesis is one of the most critical strategic subjects in organic chemistry, and this book describes advanced techniques and their applications to the industrial and laboratory synthesis of important chiral molecules. The international team of highly respected authors provide rigorous and concise reviews of their areas of expertise.
Table of Contents
Design of asymmetric synthesis.
Nonlinear effects in enantioselective synthesis - asymmetric amplification.
Chiral enolates.
Optically active B-ketosulfoxides in asymmetric synthesis.
Chiral aminals in asymmetric synthesis.
Asymmetric deprotonation reactions using enantiopure lithium amide bases.
Asymmetric Diels-Alder reactions.
Asymmetric oxidiation.
Transition metal catalysts for asymmetric reduction.
Asymmetric hydroboration.
Asymmetric hydrosilylation and hydroformylation.
Asymmetric conjugate addition reactions.
Stereoselective transformations mediated by chiral titanium complexes.
Asymmetric synthesis using enzymes and whole cells.
Asymmetric palladium catalysed coupling reactions.
Palladium allyl pi-complexes in asymmetric synthesis.
Worked examples in asymmetric synthesis design.
Stoichiometric electrophilic pi-complexes in asymmetric synthesis.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Articles
HKR Epoxides
One of the most effective and recent methods for obtaining several classes of chiral building blocks is Jacobsen’s hydrolytic kinetic resolution technique (HKR). The method provides general access to...
Aldrich ChemFiles 2005, 5.4, 17.
Keywords: Anti-inflammatory agents, Asymmetric synthesis, Building blocks, Chemfiles, Deprotonations, Ligands, Methods
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?