• USA Home
  • Z283142 - Corning® conductivity/TDS/temperature meters, benchtop Model 441
EMAIL THIS PAGE TO A FRIEND
Z283142 Aldrich
Corning® conductivity/TDS/temperature meters, benchtop Model 441
AC input 120
Purchase
Properties
feature   RS232 interface
mfr. no.   Corning
availability   not available in EU
measuring range   −5.0-105.0 temperature, resolution: 0.1 °C
  0.0001-199.9 g/L TDS, resolution: 5%
  0.01-199.9 μS conductivity, resolution: 0.01 μS
Description
General description
Features advanced software that runs a self diagnostic test upon power-up. Logs up to 10 readings and automatically recognizes up to 4 preprogrammed calibration standards. Starter kit includes electrode, electrode arm, and a selection of calibration sachets.
Legal Information
Corning is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?