• USA Home
  • Z283290 - Aldrich® TLC development chamber
EMAIL THIS PAGE TO A FRIEND
Z283290 Aldrich
Aldrich® TLC development chamber
Dual slot chamber, L × O.D. 190 mm × 25.4 mm
Purchase
Properties
L × O.D.   190 mm × 25.4 mm
Description
General description
Novel horizontal chambers permit the rapid screening of column chromatography fractions. Designed for economy and use in student laboratories. Use with 25 × 70 mm flexible plastic or glass TLC plates that may be spotted with up to 10 fractions 7 mm apart along the long edge of the plate. Uses only 2 mL solvent with elution complete in about 45 sec (using 20% EtOAc/hexanes as eluant). Greater resolution may be achieved by extending elution time. Borosilicate glass.
Performance features:
Rapid analysis; highly efficient use of TLC plate; tube wall provides a straight base for even solvent flow; very little solvent required.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z278629 Aldrich® TLC development chamber, Single slot chamber, L × O.D. 95.9 mm × 25.4 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?