• USA Home
  • Z285803 - Guide to Materials Characterization and Chemical Analysis, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z285803 Aldrich
Guide to Materials Characterization and Chemical Analysis, 2nd ed.
  •  ISBN-10 0471186333
  •  ISBN-13 978-047118633-5
Purchase
Properties
Related Categories Analytical/Chromatography, Applications, Books, Books and Software, Environmental Analysis,
publication info   J. Sibilia, John Wiley & Sons, 1996, 408 pp., hard cover
mfr. no.   Wiley
Description
General description
Written both for the novice and for the experienced scientist, this miniature encyclopedia concisely describes over one hundred materials methodologies, including evaluation, chemical analysis, and physical testing techniques. Each technique is presented in terms of its use, sample requirements, and the engineering principles behind its methodology. Real life industrial and academic applications are also described to give the reader an understanding of the significance and utilization of technique. There is also a discussion of the limitations of each technique.
Table of Contents
Molecular Spectroscopy.
Magnetic Resonance Spectroscopy.
Mass Spectrometry.
Separation Techniques.
Elemental and Chemical Analysis.
x-ray Analysis.
Microscopy.
Image Analysis.
Surface Analysis.
Thermal Analysis.
Rheology and Molecular Weight of Polymers.
Physical Properties of Particles and Polymers.
Physical Testing.
Scientific Computation.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?