• USA Home
  • Z285862 - Chromatographic Analysis of Pharmaceuticals, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z285862 Aldrich
Chromatographic Analysis of Pharmaceuticals, 2nd ed.
  •  ISBN-10 0824797760
  •  ISBN-13 978-082479776-8
Purchase
Properties
Related Categories Books and Software, Chromatography Books, Labware More...
publication info   J. Adamovics, ed., Marcel Dekker, 1996, 544 pp., hard cover
mfr. no.   Marcel Dekker
Description
General description
A new edition of the reference guide exploring the chromatographic methods used for the measurement of drugs, impurities, and excipients in pharmaceutical preparations such as tablets, ointments, and injectables. Seven chapters deal with specific chromatography topics related to regulations, sample pretreatment, gas, high-performance liquid, capillary electrophoresis, and supercritical fluid. The volume also features a 148 page applications table listing the chromatographic data of over 1300 drugs and related substances. Includes tables and graphs
Table of Contents
Regulatory Considerations for the Chromatographer, John A. Adamovics
Sample Treatment, John A. Adamovics
Planar Chromatography, John A. Adamovics and James C. Eschbach
Gas Chromatography, John A. Adamovics and James C. Eschbach
High-Performance Liquid Chromatography, John A. Adamovics and Daivd Farb
Capillary Electrophoresis, Shelley R. Rabel and John F. Stobaugh
Supercritical Fluid Chromatography of Bulk and Formulated Pharmaceuticals, James T. Stewart and Nirdosh K. Jagota
Applications, John A. Adamovics
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?