Z286001 Aldrich
Aldrich® Library of FT-IR Spectra, 2nd ed., 3-volume set
Milwaukee, WI, 1997, 5100 pp., hard cover
DOWNLOAD MSDS (PDF)
  •  ISBN-10 094163339X
  •  ISBN-13 978-094163339-0
Description
General description
This three volume set includes over 18,000 spectra categorized by chemical functionality and arranged in order of increasing structural complexity. All spectra were run in the Aldrich quality control laboratories and confirmed by other spectroscopic and chromatographic analyses. The library includes extensive cross-references by chemical name, molecular formula, CAS number, and Aldrich catalog number.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Customers Also Viewed
Aldrich<SUP>®</SUP> Library of <SUP>13</SUP>C and <SUP>1</SUP>H FT-NMR Spectra
C. Pouchert and J. Behnke, Aldrich Chemical Co., 1993, 4300 pp., hard cover
C. Pouchert, Aldrich Chemical Co., 1981, 1850 pp., hard cover
Nicolet®/Aldrich Condensed Phase Library, edition 2
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?