• USA Home
  • Z286753 - CRC Handbook of Solid State Electrochemistry
EMAIL THIS PAGE TO A FRIEND
Z286753 Aldrich
CRC Handbook of Solid State Electrochemistry
  •  ISBN-10 0849389569
  •  ISBN-13 978-084938956-6
Purchase
Properties
Related Categories Books and Software, Labware, Materials Science Books More...
publication info   , 1997, 630 pp., hard cover
mfr. no.   CRC Press
Description
General description
This book is a one stop resource treating the two main areas of solid state electrochemistry: electrochemical properties of solids such as oxides, halides, and cation conductors; and electrochemical kinetics and mechanisms of reactions occurring on solid electrolytes, including gas-phase electrocatalysis. The fundamentals are presented, including structural and defect chemistry, diffusion and transport in solids, conductivity and electrochemical reaction, and adsorption and reactions on solid surfaces. The Handbook also covers experimental methods and computer-aided interpretation of experimental results used in the field.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?