• USA Home
  • Z287350 - Analytical Instrumentation Handbook, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z287350 Aldrich
Analytical Instrumentation Handbook, 2nd ed.
  •  ISBN-10 0824794559
  •  ISBN-13 978-082479455-2
Purchase
Properties
publication info   G. Ewing, ed., Marcel Dekker, 1997, 1470 pp., hard cover
mfr. no.   Marcel Dekker
Description
General description
This Second Edition assists the novice and seasoned professional alike in systematically approaching problems with currently available tools and methods in the modern analytical chemistry domain. Covers all fields from basic theory and principles of analytical chemistry to instrumentation classification, design, and purchasing! Maintaining the expansive scope that distinguished the first edition, the Second Edition now includes new and pertinent information on x-ray methods and analysis, capillary electrophoresis, infrared and Raman technique comparisons and much more.
Table of Contents
Introduction,
The Laboratory Use of Computers,
Laboratory Balances,
Organic Elemental Analysis,
Continuous-Flow Analyzers,
Atomic Emission Spectroscopy,
Atomic Absorption and Flame Emission Spectrometry,
Ultraviolet, Visible, and Near-Infrared Spectrophotometers,
Molecular Fluorescence and Phosphorescence,
Vibrational Spectroscopy: Instrumentation for Infrared and Raman Spectroscopy,
x-ray Methods,
Photoacoustic Spectroscopy,
Techniques of Chiroptical Spectroscopy,
Nuclear Magnetic Resonanace,
Electron Paramagnetic Resonance,
x-ray Photoelectron and Auger Electron Spectroscopy
Mass Spectrometry,
Thermoanalytical Instrumentation and Applications,
Potentiometry: pH and Ion-Selective Electrodes,
Voltammetry,
Instrumentation for Stripping Analysis,
Measurement of Electrolytic Conductance,
Basic Instrumentation for Chromatography: High-Performance Liquid Chromatography,
Modern Gas Chromatographic Instrumentation,
Supercritical Fluid Chromatography Instrumentation,
Capillary Electrophoresis,
Abbreviations and Acronyms
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?