• USA Home
  • Z287601 - HPLC Methods for Pharmaceutical Analysis
EMAIL THIS PAGE TO A FRIEND
Z287601 Aldrich
HPLC Methods for Pharmaceutical Analysis
Volume 1
  •  ISBN-10 0-47118176-5
  •  ISBN-13 978-0-47118176-7
Purchase
Properties
Related Categories Books and Software, Chromatography Books, Labware More...
publication info   G. Lunn and N. Schmuff, John Wiley & Sons, 1997, 1632 pp., hard cover
mfr. no.   Wiley
Description
General description
Lets you target the specific information you need quickly and easily -so you can stay in the lab and out of the library. This book gives you fast, hands-on access to the most significant literature available on HPLC for the analysis of over 150 of the most frequently prescribed pharmaceutical compounds. Combining full descriptions of procedures with specially annotated references, this book helps you to identify the techniques that are relevant to your individual project needs -eliminating hours of tedious, time-consuming library research. Analytical chemists and researchers will find a battery of useful features that make this practical resource a welcome addition to their reference library: Techniques for each compound are described in detail -enabling replication of a procedure without reference to the original publication.
Provides fast, hands-on access to thousands of abstracts from the current literature on HPLC methods for pharmaceutical analysis.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?