• USA Home
  • Z287636 - Spectra Interpretation of Organic Compounds
EMAIL THIS PAGE TO A FRIEND
Z287636 Aldrich
Spectra Interpretation of Organic Compounds
  •  ISBN-10 3527288260
  •  ISBN-13 978-352728826-7
Purchase
Properties
Related Categories Books and Software, Labware, Spectroscopy Books More...
publication info   E. Pretsch and J. Clerc, John Wiley & Sons, Ltd, 1997, 190 pp., soft cover/CD-ROM
mfr. no.   Wiley
Description
General description
A unique advanced textbook on spectroscopy. This interactive tutorial presents text, software and data in a state-of-the-art introduction to the interpretation of 13C- and 1H-nuclear magnetic resonance, infrared, mass and UV/VIS spectra. Designed as a hands-on guide, the newcomer or student learns not only by reading but by experimenting, using the powerful software tools and data provided on the accompanying CD-ROM. The software, based on the outstanding SpecTool product, enables you to learn how to interpret molecular spectra correctly, rapidly and easily. Moreover, you can check your progress by working through the examples embedded in this self-study course that demonstrate how to identify an organic compound and to elucidate its structure. All the material and software presented are the essence of the two authors´ longstanding teaching experience.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?