• USA Home
  • Z287776 - Introduction to X-ray Powder Diffractometry
EMAIL THIS PAGE TO A FRIEND
Z287776 Aldrich
Introduction to X-ray Powder Diffractometry
  •  ISBN-10 0471513393
  •  ISBN-13 978-047151339-1
Purchase
Properties
Related Categories Analytical Chemistry Books, Books and Software, Labware More...
publication info   R. Jenkins, John Wiley & Sons, Ltd, 1996, 432 pp., hard cover
mfr. no.   Wiley
Description
General description
Solid materials leave distinct patterns similar to fingerprints if they are x-rayed. x-ray powder diffractometry is an established technique used to fingerprint and identify samples such as moon rocks or pharmaceutical ingredients. This book provides an up-to-date, thorough introduction to the field, with clear, concise descriptions of both theory and practice.
Table of Contents
Characteristics of X-Radiation.
The Crystalline State.
Diffraction Theory.
Sources for the Generation of X-Radiation.
Detectors and Detection Electronics.
Production of Monochromatic Radiation.
Instruments for the Measurement of Powder Patterns.
Alignment and Maintenance of Powder Diffractometers.
Specimen Preparation.
Acquisition of Diffraction Data.
Reduction of Data from Automated Powder Diffractometers.
Qualitative Analysis.
Quantitative Analysis.
Appendices.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?