EMAIL THIS PAGE TO A FRIEND
Z287989 Aldrich
Single input meter
Type-K
Purchase
Properties
Related Categories Barnant, Barnant Temperature Measurement, Labware, Manufacturer Browser, Temperature,
mfr. no.   Barnant
L × W × D   158 mm × 84 mm × 30 mm
Description
General description
Meters feature a Hold function, selectable °C or °F, field calibration, and a low battery indicator. Includes builtin tilt stand and two AA batteries. Order thermocouples separately.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?