• USA Home
  • Z289132 - Thermocouple probes small diameter
EMAIL THIS PAGE TO A FRIEND
Z289132 Aldrich
Thermocouple probes small diameter
Small diameter, Type-K
Purchase
Properties
Related Categories Barnant, Barnant Temperature Measurement, Labware, Manufacturer Browser, Temperature,
mfr. no.   Barnant
parameter   -250-816 °C temp. range (-418-1500 °F)
L × diam.   4 in. × 1/16 in.
Description
General description
Quality probes compatible with all thermocouple meters and controllers that use miniconnectors. Probe is 316 stainless steel, small diameter, 4 × 0.062, with 5 1/2 in. long glass-filled nylon handles and 5ft. coiled cable with miniconnectors.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?