• USA Home
  • Z289469 - Practice of Medicinal Chemistry
EMAIL THIS PAGE TO A FRIEND
Z289469 Aldrich
Practice of Medicinal Chemistry
  •  ISBN-10 0127446400
  •  ISBN-13 978-012744640-0
Purchase
Properties
publication info   C. Wermuth, ed., Academic Press, 1996, hard cover
mfr. no.   Elsevier
Description
General description
This book collects in one single volume, the practical aspects of Medicinal Chemistry, seen from a chemical point of view, including the wealth of information which chemists accumulate over a career, but generally is never organized and presented in a coherent form in print. Emphasis is given to how medicinal chemists conduct their search for, and design of, new drug entities. In contrast to other books on the market, it focuses on the chemistry, rather than pharmacological concepts or description of the various therapeutic classes of drugs. It should become a standard reference on the tools available to medicinal chemists when designing new drugs.
Table of Contents
General Aspects of Medicinal Chemistry
Drug Targets and Lead Compound Discovery Strategies
Primary Exploration of Structure Activity Relationships
Spatial Organization, Receptor Mapping and Molecular Modeling
Chemical Modifications Influencing the Pharmacokinetic Properties
Pharmaceutical and Chemical Formulation Problems
Development of New Drugs: Legal and Economic Aspects
Subject Index
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?