• USA Home
  • Z289507 - Barnant thermocouple probe
EMAIL THIS PAGE TO A FRIEND
Z289507 Aldrich
Barnant thermocouple probe
Flexible high-temp, Type-K
Purchase
Properties
Related Categories Barnant, Barnant Temperature Measurement, Labware, Manufacturer Browser, Temperature,
label   C
mfr. no.   Barnant
parameter   1100 °C temp. limit (≤2012 °F)
L × W   12 in. × 0.04 in.
Description
General description
Probe is MgO filled Inconel with a 3ft. straight cable with miniconnectors, no handle.
Quality probes compatible with all thermocouple meters and controllers that use miniconnectors. Note: Temperature range on some probes is limited due to materials of construction.
• 316 SS probes
• 51/2 in. L glass-filled nylon handles
• 5 ft coiled cables with miniconnectors
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?