• USA Home
  • Z291064 - Hamilton® 700 series syringe for Hewlett-Packard 7673A autosampler
EMAIL THIS PAGE TO A FRIEND
Z291064 Aldrich
Hamilton® 700 series syringe for Hewlett-Packard 7673A autosampler
Model 75 ASN, capacity 5 μL
Purchase
Properties
Related Categories 700 Series, Hamilton syringes, Labware, Syringes and Needles
description   HP®
point style needle
  cemented needle
mfr. no.   Model 75 ASN
capacity   5 μL
needle L × I.D.   1.71 in. × 0.13 mm
needle size × O.D.   23 S gauge × 0.64 mm
Description
Legal Information
HP is a registered trademark of Hewlett-Packard Corp.
Hamilton is a registered trademark of Hamilton Co.
Hewlett-Packard is a trademark of Hewlett-Packard Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
21311 Hamilton® Microliter syringe, cemented needle, 75 ASN, volume 5 μL, needle size 23s ga (cone tip), needle L 43 mm (1.71 in.), pkg of 1 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?