• USA Home
  • Z292060 - Septa for aluminum caps
EMAIL THIS PAGE TO A FRIEND
Z292060 Aldrich
Septa for aluminum caps
Clear FEP/silicone septa
Purchase
Properties
description   For autosampler headspace analysis
packaging   pkg of 100 ea
  pkg of 1000 ea
diam.   20 mm
Description
General description
20mm caps for the autosampler vials listed above.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
27016 Aluminum crimp seal (without septum), silver aluminum, tear-away, diam. 20 mm, pkg of 100 ea
27200 Aluminum crimp seal (without septum), silver aluminum, pressure release, diam. × opening 20 mm × 8 mm, pkg of × 100 ea
27227-U Aluminum crimp seal (without septum), silver aluminum, removable center, diam. 20 mm, pkg of 100 ea
27230-U Aluminum crimp seal (without septum), silver aluminum, open center, diam. × opening 20 mm × 9.5 mm, pkg of 100 ea
33183 Hand Decapper, for use with 20mm crimp seals, pkg of 1 ea
33280-U Crimpers, Hand crimper, for use with 20mm seals, pkg of 1 ea
22316-U Hand crimper, adjustable, Hand crimper, adjustable, for use with 20 mm crimp seals, pkg of 1 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?