• USA Home
  • Z293008 - TLC Glass Plates from EMD/Millipore
EMAIL THIS PAGE TO A FRIEND
Z293008 Aldrich
TLC Glass Plates from EMD/Millipore
L × W 5 cm × 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent indicator
  •  eCl@ss 32110504
Purchase
Properties
Related Categories Analytical/Chromatography, Chromatography, Chromatography, TLC Plates, EMD Millipore TLC Plates, Glass TLC Plates: Silica matrix,
material   glass support
  silica gel 60 matrix
feature   binder Polymeric
  fluorescent indicator
packaging   pkg of 100 ea
mfr. no.   EMD Millipore, 1.05714.0001
application(s)   thin layer chromatography (TLC): suitable
L × W   5 cm × 20 cm
layer thickness   250 μm
particle size   10-12 μm
pore size   60 Å
Description
Other Notes
With fluorescent indicator on glass
Price and Availability
Pharma QC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?