• USA Home
  • Z300322 - Tyvek® disposable footwear
EMAIL THIS PAGE TO A FRIEND
Z300322 Aldrich
Tyvek® disposable footwear
Overboots, size L
Purchase
Properties
Related Categories Footwear, Labware, Safety Clothing, Safety Equipment
size   L
mfr. no.   Tyvek
availability   not available in USA and Canada
Description
General description
Manufactured from Tyvek with elasticated tops, this footwear is low linting, biologically inert, has good particle barrier properties, some resistance to permeation and penetration of many chemicals and might be used in controlled, clean or hazardous environments.
Legal Information
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?