• USA Home
  • Z300365 - Tyvek® disposable aprons
EMAIL THIS PAGE TO A FRIEND
Z300365 Aldrich
Tyvek® disposable aprons
Tyvek, PE coated
Purchase
Properties
Related Categories Aprons, Oversleeves and Hoods, Labware, Safety Clothing, Safety Equipment
mfr. no.   Tyvek
availability   not available in USA and Canada
Description
General description
Choice of Tyvek for antistatic, low linting and particle barrier properties combined with some resistance to permeation and penetration of many chemicals, or PE coated Tyvek for additional liquid and chemical splash protection. Aprons have halter neck with waist ties. Sleeves are elasticated at both ends.
Legal Information
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?