• USA Home
  • Z301000 - Quickfit® Reduction Adapters
EMAIL THIS PAGE TO A FRIEND
Z301000 Aldrich
Quickfit® Reduction Adapters
top female Joint: ST/NS 24/29, bottom male Joint: ST/NS 50/42
Purchase
Properties
Related Categories Enlarging/Reducing Adapters, Glassware, Glassware Adapters, Labware, Quickfit reduction More...
Joint   female Joint: ST/NS top24/29
  male Joint: ST/NS bottom50/42
mfr. no.   Quickfit DA 38
availability   not available in USA and Canada
Description
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?