• USA Home
  • Z301345 - Quickfit® plain bends
EMAIL THIS PAGE TO A FRIEND
Z301345 Aldrich
Quickfit® plain bends
Joint: ST/NS 34/35
Purchase
Properties
Related Categories Bent Glass Adapters, Glassware, Glassware Adapters, Labware
Joint   Joint: ST/NS34/35
mfr. no.   Quickfit RA 9/55
availability   not available in USA and Canada
  not available in USA and Canada
Description
General description
Socket to cone. Generally used as distilling adapters.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?