• USA Home
  • Z301604 - Marigold® Featherlite natural rubber gloves
EMAIL THIS PAGE TO A FRIEND
Z301604 Aldrich
Marigold® Featherlite natural rubber gloves
size S
Purchase
Properties
Related Categories Disposable Gloves, Disposable Latex Gloves, Featherlite natural rubber gloves, Gloves, Labware,
size   S
feature   CE compliant
packaging   pkg of 100 ea
mfr. no.   Marigold
availability   not available in USA and Canada
L × thickness   240 mm × 0.20 mm , double wall
Description
General description
Supplied in dispenser style boxes, these powdered easy pull-on disposable gloves are ambidexterous. Tear resistant cuff beading, textured surface and high sensitivity make Featherlite good general purpose gloves for laboratory work.
Legal Information
Marigold is a registered trademark of Comasec SAS
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?