• USA Home
  • Z301647 - Marigold® Suretouch vinyl gloves
EMAIL THIS PAGE TO A FRIEND
Z301647 Aldrich
Marigold® Suretouch vinyl gloves
size M
Purchase
Properties
Related Categories Disposable Gloves, Disposable Vinyl Gloves, Gloves, Labware, Manufacturer Browser,
size   M
feature   CE compliant
packaging   pkg of 100 gloves
mfr. no.   Marigold
availability   not available in USA and Canada
L × thickness   240 mm × 0.20 mm , double wall
Description
General description
Easy pull-on, tear resistant, disposable vinyl gloves packed in dispenser style boxes offering the wearer high sensitivity when in use. Vinyl gloves offer protection against minimal risks only.
Legal Information
Marigold is a registered trademark of Comasec SAS
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?