• USA Home
  • Z301701 - Marigold® Suretech vinyl gloves
EMAIL THIS PAGE TO A FRIEND
Z301701 Aldrich
Marigold® Suretech vinyl gloves
size L, standard length
Purchase
Properties
Related Categories Disposable Gloves, Disposable Vinyl Gloves, Gloves, Labware, Manufacturer Browser,
size   L
feature   CE compliant
  standard length
packaging   pkg of 1000 ea
mfr. no.   Marigold
availability   not available in USA and Canada
L × thickness   240 mm × 0.26 mm
Description
General description
Available in standard or 25% longer length, these ambidextrous gloves are ideal for use in laboratory, pharmaceutical or electronic environments. Double bagged, powder-free and with low Chloride, Sulfur and Sodium ion levels, they combine minimum particulate contamination with good sensitivity, grip and tear resistance.
Legal Information
Marigold is a registered trademark of Comasec SAS
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?