• USA Home
  • Z301930 - Marigold® blue nitrile gloves
EMAIL THIS PAGE TO A FRIEND
Z301930 Aldrich
Marigold® blue nitrile gloves
size S
Purchase
Properties
Related Categories Blue nitrile gloves, Gloves, Heavyweight Gloves, Labware, Manufacturer Browser,
material   blue nitrile
size   S
feature   CE compliant
packaging   pkg of 12 pair
mfr. no.   Marigold
availability   not available in USA and Canada
L × thickness   330 mm × 0.84 mm
Description
General description
Manufactured from acrylonitrile rubber, these gloves offer good resistance to many solvents and oil based chemicals. They are flock lined for comfort and have an open cuff which allows ventilation, keeping the hands cool. Strong and flexible for comfort in extended use and with a patterned finish for good grip.
Legal Information
Marigold is a registered trademark of Comasec SAS
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?