• USA Home
  • Z302147 - Tyvek® Pro.Tech total body coveralls
EMAIL THIS PAGE TO A FRIEND
Z302147 Aldrich
Tyvek® Pro.Tech total body coveralls
size S
Purchase
Properties
Related Categories Coveralls, Labware, Safety Clothing, Safety Equipment
size   S
mfr. no.   Tyvek
availability   not available in USA and Canada
Description
General description
With full length zipper, elasticated wrist holes, integral elasticated hood and overshoes for protection in controlled, clean environments. Provides CE type 5 and Type 6 (particle and limited splash) protection.
Legal Information
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z740475 MICROGARD 2000 coveralls for protection from infective agent, size S (boot ends with anti-slip soles)
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?