• USA Home
  • Z302228 - Tyvek® Pro.Tech coveralls
EMAIL THIS PAGE TO A FRIEND
Z302228 Aldrich
Tyvek® Pro.Tech coveralls
size M
Purchase
Properties
Related Categories Coveralls, Labware, Safety Clothing, Safety Equipment
size   M
mfr. no.   Tyvek
availability   not available in USA and Canada
Description
General description
With integral hood, full length zipper and elasticated wrist and ankle holes. Zipper has covering flap and seams are taped for extra protection. Provides CE Type 5 and Type 6 (particle and limited splash) protection.
Legal Information
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?