• USA Home
  • Z303461 - Quickfit® two-neck, round-bottom flasks

EMAIL THIS PAGE TO A FRIEND
Z303461 Aldrich

Quickfit® two-neck, round-bottom flasks

capacity 1,000 mL, center Joint: ST/NS 24/29

Purchase

Properties

Related Categories Flasks, Labware, Multi-neck Flasks, Round-bottom Flasks
material   borosilicate glass
  flask round bottom
  two-neck (One vertical central neck and one angled side neck)
Joint   center Joint: ST/NS24/29
  side Joint ST/NS19/26
sterility   non-sterile
feature   closure type Standard Taper Joint
packaging   pack of 1
mfr. no.   Quickfit
availability   not available in USA and Canada
H   180 mm
capacity   1,000 mL

Description

General description

One vertical central neck and one angled side neck

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?