• USA Home
  • Z303593 - Quickfit® three-neck round-bottom flasks with angled necks

EMAIL THIS PAGE TO A FRIEND
Z303593 Aldrich

Quickfit® three-neck round-bottom flasks with angled necks

capacity 1,000 mL, center Joint: ST/NS 29/32, side Joint: ST/NS 29/32

Purchase

Properties

Related Categories Flasks, Labware, Multi-neck Flasks, Round-bottom Flasks
material   borosilicate glass
  flask round bottom
  three angled neck
Joint   center Joint: ST/NS29/32
  side Joint: ST/NS29/32
sterility   non-sterile
feature   closure type Standard Taper Joint
packaging   pack of 1
mfr. no.   Quickfit
availability   not available in USA and Canada
H   183 mm
capacity   1,000 mL

Description

General description

With three necks fitted with Quickfit, precision-ground sockets. Axes of all three joints meet at bottom of flask.
• One vertical neck
• Two angled side necks

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?