• USA Home
  • Z303739 - Quickfit® three-neck round-bottom flasks with vertical necks
EMAIL THIS PAGE TO A FRIEND
Z303739 Aldrich
Quickfit® three-neck round-bottom flasks with vertical necks
capacity 2,000 mL, center Joint: ST/NS 45/40
Purchase
Properties
Related Categories Flasks, Labware, Multi-neck Flasks, Round-bottom Flasks
material   borosilicate glass
  flask round bottom
  three-neck (Vertical)
Joint   center Joint: ST/NS45/40
  side Joint ST/NS29/32
sterility   non-sterile
feature   closure type Standard Taper Joint
packaging   pack of 1
mfr. no.   Quickfit
availability   not available in ROW (Available only in EU countries)
  not available in USA and Canada
  not available in USA and Canada
H   233 mm
capacity   2,000 mL
Description
General description
• One vertical neck
• Two parallel necks
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?