• USA Home
  • Z303836 - Quickfit® wide-neck reaction flask

EMAIL THIS PAGE TO A FRIEND
Z303836 Aldrich

Quickfit® wide-neck reaction flask

capacity 4,000 mL, Spherical

Purchase

Properties

Related Categories Flasks, Labware, Reaction Flasks More...
material   borosilicate glass
  wide-neck
sterility   non-sterile
feature   closure type Lid
  Spherical
packaging   pack of 1
mfr. no.   Quickfit, FR 4LF
availability   not available in USA and Canada
capacity   4,000 mL

Description

General description

Reaction flasks have a flat flange so that lids can be quickly and easily detached. Flanges have 100 mm bore for easy addition or removal of starting materials and products, as well as for simple cleaning. Z653322 has 75mm bore. Order lids and clips separately.

NOTE: Z338052 is cylindrical.

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?