• USA Home
  • Z303968 - Quickfit® jacketed reaction flasks
EMAIL THIS PAGE TO A FRIEND
Z303968 Aldrich
Quickfit® jacketed reaction flasks
capacity 1,000 mL
Purchase
Properties
Related Categories Flasks, Labware, Reaction Flasks More...
material   borosilicate glass
sterility   non-sterile
packaging   pack of 1
mfr. no.   Quickfit, JRV 1L
availability   not available in USA and Canada
capacity   1,000 mL
Description
General description
Water jacket for heating or cooling flask contents. For lids and clamp please order separately.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?