• USA Home
  • Z304409 - Quickfit® dropping funnel with GP Rotaflo® stopcock
EMAIL THIS PAGE TO A FRIEND
Z304409 Aldrich
Quickfit® dropping funnel with GP Rotaflo® stopcock
capacity 50 mL, Joint: ST/NS 14/23
Purchase
Properties
Related Categories Addition Funnels, Addition/Dropping, Funnels, Glass Funnels, Glassware,
Joint   Joint: ST/NS14/23
mfr. no.   Quickfit, D 31/1/50
availability   not available in USA and Canada
capacity   50 mL
Description
General description
Cylindrical with ground-glass joint, GP Rotaflo stopcocks and the stem in line with cone
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Rotaflo is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?