• USA Home
  • Z304557 - Quickfit® pressure equalising dropping funnel
EMAIL THIS PAGE TO A FRIEND
Z304557 Aldrich
Quickfit® pressure equalising dropping funnel
capacity 500 mL, Cylindrical, male Joint: ST/NS 19/26, female Joint: ST/NS 24/29
Purchase
Properties
Related Categories Addition Funnels, Addition/Dropping, Funnels, Glass Funnels, Glassware,
Joint   female Joint: ST/NS24/29
  male Joint: ST/NS19/26
feature   Cylindrical
mfr. no.   Quickfit, DEC 500/32
availability   not available in USA and Canada
capacity   500 mL
Description
General description
With GP Rotaflo stopcocks
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?