• USA Home
  • Z304670 - Quickfit® ground-glass stoppers
EMAIL THIS PAGE TO A FRIEND
Z304670 Aldrich
Quickfit® ground-glass stoppers
Joint: ST/NS 55/44
Purchase
Properties
Related Categories Glass Stoppers, Labware, Stoppers More...
Joint   Joint: ST/NS55/44
mfr. no.   Quickfit, SB 55
availability   not available in USA and Canada
Description
General description
• Ideally suited for use with Quickfit® jointedware and other glassware having standard ground glass sockets
• All Quickfit® stoppers conform to the standard tapers as outlined in ISO 383
• Borosilicate glass for chemical durability and high temperature performance
• Hollow-blown construction to reduce weight
• Hexagonal head prevents the stopper from rolling and provides a good grip
• Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?