• USA Home
  • Z304719 - Quickfit® enclosed-scale thermometers
EMAIL THIS PAGE TO A FRIEND
Z304719 Aldrich
Quickfit® enclosed-scale thermometers
Joint: ST/NS 14/23, −10-110 °C temperature
Purchase
Properties
Related Categories Labware, Temperature, Thermometers More...
Joint   Joint: ST/NS14/23
mfr. no.   Quickfit, MF 31/1
availability   not available in USA and Canada
measuring range   −10-110 °C temperature
Description
General description
With integral cone for use with jointed apparatus
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?