• USA Home
  • Z304727 - Quickfit® enclosed-scale thermometers
EMAIL THIS PAGE TO A FRIEND
Z304727 Aldrich
Quickfit® enclosed-scale thermometers
Joint: ST/NS 14/23, −10-250 °C temperature
Purchase
Properties
Related Categories Labware, Temperature, Thermometers More...
Joint   Joint: ST/NS14/23
mfr. no.   Quickfit, MF 31/2
availability   not available in USA and Canada
measuring range   −10-250 °C temperature
Description
General description
With integral cone for use with jointed apparatus
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?