• USA Home
  • Z306584 - Volac® volumetric flask
EMAIL THIS PAGE TO A FRIEND
Z306584 Aldrich
Volac® volumetric flask
2,000 mL volume, accuracy: 0.06 mL, certificate
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Poulten Graf, Pyrex Volumetric Flasks, Volac volumetric flask,
stopper size size   20‑24
feature   certificate
  class A
mfr. no.   Poulton & Graf
availability   not available in USA and Canada
measuring range   2,000 mL volume, accuracy: 0.06 mL
Description
General description
Conform to BS1792 and ISO1042. Each Volac flask is individually calibrated and printed, tested for accuracy, and numbered for full traceability, regardless whether Works Correction Certificate of Accuracy is provided. Borosilicate glass, permanent markings, and tooled ground neck fitted with PP stopper.
Legal Information
Volac is a registered trademark of Poulten & Graf Ltd.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?