• USA Home
  • Z307203 - Volac® volumetric pipette
EMAIL THIS PAGE TO A FRIEND
Z307203 Aldrich
Volac® volumetric pipette
100 mL volume, accuracy: 0.08 mL
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Poulten Graf, Volac volumetric pipette, Volumetric,
material   borosilicate glass
feature   class AS
  (Conforms to BS ISO648, batch tested)
mfr. no.   Poulton & Graf
availability   not available in USA and Canada
measuring range   100 mL volume, accuracy: 0.08 mL
capacity   100 mL
color   yellow
Description
General description
Color-coded for easy recognition and calibrated to deliver (TD, EX). "Tufftip" treated tips resist damage on knocking.
Legal Information
Volac is a registered trademark of Poulten & Graf Ltd.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?