• USA Home
  • Z312428 - Variomag MONOTHERM hotplate stirrer

EMAIL THIS PAGE TO A FRIEND
Z312428 Aldrich

Variomag MONOTHERM hotplate stirrer

gray

Purchase

Properties

Related Categories Heating, Hotplate Stirrers, Labware More...
AC input   230 V
material   gray
grade   CE Compliant
sterility   non-sterile
mfr. no.   H+P Labortechnik
  Hewlett-Packard
availability   not available in USA and Canada
L × W × H   160 mm × 295 mm × 60 mm
color   none
matrix active group   Non-treated surface treatment

Description

General description

Magnetic stirrers with 500 W heaters for heating up to 300 °C. Colored aluminum housing.
Unlike conventional magnetic stirrers with electric motors spinning permanent magnets, VARIOMAG stirrers have no moving parts. VARIOMAG stirrers have stationary magnetic coils through which current is varied to produce a rotating magnetic field that drives the magnetic stir bar. As a result, VARIOMAG stirrers have a flat design with no wear, no maintenance and high precision speed control.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?