• USA Home
  • Z338222 - Handbook of Reagents for Organic Synthesis: Oxidizing and Reducing Agents
EMAIL THIS PAGE TO A FRIEND
Z338222 Aldrich
Handbook of Reagents for Organic Synthesis: Oxidizing and Reducing Agents
S. Burke, ed., John Wiley & Sons, Ltd, 1999, 564 pp., hard cover
  •  ISBN-10 0471979260
  •  ISBN-13 978-047197926-5
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   S. Burke, ed., John Wiley & Sons, Ltd, 1999, 564 pp., hard cover
mfr. no.   Wiley
Description
General description
One of four self-contained handbooks derived from the renowned Encyclopedia of Reagents for Organic Synthesis, this volume focuses on oxidizing and reducing agents for organic synthesis. It follows the same format as the Encyclopedia, including an introduction and an alphabetical arrangement of the reagents. It also provides physical and detailed chemical data for every reagent, helping chemists in diverse disciplines understand what the reagents are used for and how to work safely with them.
Table of Contents
Partial Table of Contents:
Aluminum Amalgam.
Aluminum Hydride.
Borane-Tetrahydrofuran.
Bromine.
Chromium(II) Chloride.
Chromium(VI) Oxide.
Hydrogen Sulfide.
Iodosylbenzene.
Lithium-Ethylamine.
Lithium Naphthalenide.
Manganese Dioxide.
Mercury(II) Oxide.
Palladium on Carbon.
Palladium(II) Hydroxide on Carbon.
Peracetic Acid.
Rhodium on Alumina.
Ruthenium Catalysts.
Ruthenium(VIII) Oxide.
Sodium-Alcohol.
Sodium Amalgam.
Sodium-Ammonia.
Sulfur.
Thallium(III) Nitrate Trihydrate.
Thallium(III) Trifluoroacetate.
Thexylborane.
Zinc.
Zinc Borohydride.
Zinc/Copper Couple.
List of Contributors.
Indexes.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?