• USA Home
  • Z400351 - Biodisposer portable sharps collection containers
EMAIL THIS PAGE TO A FRIEND
Z400351 Aldrich
Biodisposer portable sharps collection containers
capacity 1 L
Purchase
Properties
Related Categories Disposal Containers, Labware, Safety Equipment, Sharps Disposal Contaners
L × W × H   3 1/2 in. × 3 1/2 in. × 6 in.
capacity   1 L
Description
General description
These containers ensure safe and convenient disposal of syringe needles and other sharp hazardous waste. Safely resheaths or unwinds needles after use. Stackable red PP bases with white snap-on covers. Safety labeled.
Legal Information
Biodisposer is a trademark of Simport
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?