• USA Home
  • Z400750 - Chromatography funnel
EMAIL THIS PAGE TO A FRIEND
Z400750 Aldrich
Chromatography funnel
Set of one each of five funnels
Purchase
Properties
Related Categories Glass Chromatography Columns, Glassware, Gravity, Labware
material   polypropylene (Unbreakable)
packaging   pkg of 5 ea
Description
General description
Use for solvent addition in flash chromatography. Designed to not disturb the top layer of the column; a perforated stem with a closed-end permits solvent to pour down column walls.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?