| Related Categories | Labware, MSA, Manufacturer Browser, Respirators, Reusable Respirators, |
| size | S |
| mfr. no. | MSA, 805414 |
| availability | not available in EU |
• Economical and low-maintenance
• Light weight
• Speaking diaphragm for clear communications
• Black, super soft Hycar rubber facepiece for comfort and durability
• Wrap-around flexible lens offering excellent fit and field of vision
• Nose cup to reduce fogging
Supplied without cartridges. Order Advantage 1000 Super cartridges separately. NIOSH/MSHA approved.
Advantage is a registered trademark of Mine Safety Appliances Co.
Customers Also Viewed
GME type, for Advantage® Series respirator
GME/P100 type, for Advantage® Series respirator