• USA Home
  • Z402184 - Infrared and Raman Spectra of Inorganic and Coordination Compounds, Part A: Theory and Applications in Inorganic Chemistry, 5th ed.
EMAIL THIS PAGE TO A FRIEND
Z402184 Aldrich
Infrared and Raman Spectra of Inorganic and Coordination Compounds, Part A: Theory and Applications in Inorganic Chemistry, 5th ed.
  •  ISBN-10 0471163945
  •  ISBN-13 978-047116394-7
Purchase
Properties
publication info   K. Nakamoto, John Wiley & Sons, Ltd, 1997, hard cover
mfr. no.   Wiley
Description
General description
This book, along with its companion volume, is a thoroughly revised and expanded edition of a best-seller. Completely self-contained, Part A presents a clear introduction to infrared and raman spectroscopy--two techniques used to fingerprint and identify chemical substances. Beginning with fundamental theory, it goes on to present some basic applications in inorganic chemistry.
Table of Contents
PARTIAL TABLE OF CONTENTS:
THEORY OF NORMAL VIBRATIONS.
Origin of Molecular Spectra.
Vibration of a Diatomic Molecule.
Normal Coordinates and Normal Vibrations.
Symmetry of Normal Vibrations and Selection Rules.
The Number of Normal Vibrations for Each Species.
Selection Rules for Infrared and Raman Spectra.
Utilization of Symmetry Properties.
Potential Fields and Force Constants.
Solution of the Secular Equation.
Vibrational Frequencies of Isotopic Molecules.
Depolarization of Raman Lines.
Intensity of Raman Scattering.
Resonance Raman Spectra.
Lattice Vibrations.
APPLICATIONS IN INORGANIC CHEMISTRY.
Diatomic Molecules.
Triatomic Molecules.
Planar Four-Atom Molecules.
Tetrahedral and Square-Planar Five-Atom Molecules.
Metal Cluster Compounds.
Compounds of Carbon.
Compounds of Nitrogen.
Compounds of Sulfur and Selenium.
Appendices.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?