• USA Home
  • Z402230 - Oxford Chemistry Primers
EMAIL THIS PAGE TO A FRIEND
Z402230 Aldrich
Oxford Chemistry Primers
No. 50: Heat Transfer
  •  ISBN-10 0198562977
  •  ISBN-13 978-019856297-9
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   R. Winterton, Oxford University Press, 1997, 85 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
This primer follows a classical approach to the study of heat transfer with main chapters on conduction, forced convection, natural convection and radiation. Practical problems, however, do not normally divide neatly into purely conduction problems, purely forced convection and so on, although texts often assume this. To overcome this barrier, the author demonstrates in examples and problems that more than one aspect of heat transfer may be needed to solve a single practical problem. The book covers the essential material of a text twice its length, making it easier for the reader to find and understand key principles. It will be of particular interest to undergraduate students in engineering, especially in mechanical and chemical engineering, and to professional engineers working on heat transfer.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?