| Related Categories | Buchi, Buchi Glass Rotary Evaporator Parts, Buchi Parts List, Condensers for Buchi Rotary Evaporators, Dry-Ice (Buchi) High-Capacity Condensers, |
| material | plastic-coated glass |
| mfr. no. | Buchi, 032304 |
Comes with KD-22 vacuum seal.
Condensers for R200/205 and R210/215 come with hose barbs and drain sleeve. Replacement condensers do not include stopcocks.
Büchi is a registered trademark of Büchi Labortechnik AG
Rotavapor is a registered trademark of Büchi Labortechnik AG