• USA Home
  • Z403601 - IKA® supports for stirring hot plates
EMAIL THIS PAGE TO A FRIEND
Z403601 Aldrich
IKA® supports for stirring hot plates
R 380 adjustable stand support, stainless steel, 1/cs
Purchase
Properties
Related Categories Heating, Hotplate Stirrers, IKA, IKA Heating Equipment and Accessories, IKA Support for Stirring Hotplate, Adjustable Stand Clamp,
material   stainless steel
packaging   case of 1
  pack of 1
mfr. no.   IKA 2636700
Description
General description
Attaches to the side groove of the IKA® stirring hot plate allowing adjustment to be fixed at any position. Several support rods can be attached to stirrer simultaneouslty. Order support rod Z40,356-3 separately.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?