• USA Home
  • Z403644 - IKA® Color Squid IKAMAG magnetic plate stirrers
EMAIL THIS PAGE TO A FRIEND
Z403644 Aldrich
IKA® Color Squid IKAMAG magnetic plate stirrers
small magnetic stirrer, Number One top plate design, 1/cs
Synonym: magnetic plate stirrers, magnetic stirrers, stirrer, stirrers
Purchase
Properties
Related Categories IKA, IKA Squid Stirrers, IKA Stirrers, Labware, Magnetic Stirrers,
material   Number One top plate design
packaging   case of 1
  pack of 1
mfr. no.   IKA 2669917
Description
General description
IKA Color Squid IKAMAG
Small magnetic stirrers in various motifs. Recyclable materials with very good chemical resistance provided by glass top and synthetic lower part made of HYTREL.
• Electronically controlled motor
• Speed range: 0-1500 rpm
• Stirring volume: 800 mL (water)
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
IKAMAG is a trademark of IKA-Werke GmbH & Co. KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?